ENA048303803 Enamine

2-mercapto-4,6-dimethylnicotinamide

  • MDL number MFCD00473618
  • Cas Number : 79927-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...170
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11478

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11478
Availability details for
Quantity
  • Availability
  • Add to Cart