ENA048303804 Enamine

ethyl 5-cyano-6-mercapto-2-methylnicotinate

  • MDL number MFCD00173692
  • Cas Number : 113858-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11479

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11479
Availability details for
Quantity
  • Availability
  • Add to Cart