ENA048303811 Enamine

3-{[(4-isopropylphenyl)sulfonyl]amino}propanoic acid

  • MDL number MFCD06655215
  • Cas Number : 857041-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...205
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11003

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11003
Availability details for
Quantity
  • Availability
  • Add to Cart