ENA048303817 Enamine

2-[(4-tert-butylbenzoyl)amino]propanoic acid

  • MDL number MFCD02323126
  • Cas Number : 726174-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08398

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08398
Availability details for
Quantity
  • Availability
  • Add to Cart