ENA048303824 Enamine

2-[(4-bromobenzoyl)amino]propanoic acid

  • MDL number MFCD00686879
  • Cas Number : 183559-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08413

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08413
Availability details for
Quantity
  • Availability
  • Add to Cart