ENA048303833 Enamine

2-(2,3-dihydro-1,4-benzodioxin-6-yl)pyrrolidine

  • MDL number MFCD04966886
  • Cas Number : 524674-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11186

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11186
Availability details for
Quantity
  • Availability
  • Add to Cart