ENA048303836 Enamine

1-(2-chloroacetyl)imidazolidin-2-one

  • MDL number MFCD06363490
  • Cas Number : 57013-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-91
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11188
Availability details for
Quantity
  • Availability
  • Add to Cart