ENA048303839 Enamine

2-(4-nitrobenzenesulfonyl)acetic acid

  • MDL number MFCD03983033
  • Cas Number : 3937-94-8
  • Molecular Weight : 245.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11514
Availability details for
Quantity
  • Availability
  • Add to Cart