ENA048303845 Enamine

3-{[(3,4-difluorophenyl)sulfonyl]amino}propanoic acid

  • MDL number MFCD06357896
  • Cas Number : 746655-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11015

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11015
Availability details for
Quantity
  • Availability
  • Add to Cart