ENA048303847 Enamine

1-[(3,4-difluorophenyl)sulfonyl]piperidine-4-carboxylic acid

  • MDL number MFCD01569393
  • Cas Number : 716361-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11016

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11016
Availability details for
Quantity
  • Availability
  • Add to Cart