ENA048303849 Enamine

1-phenylmethanesulfonylpiperazine

  • MDL number MFCD03444529
  • Cas Number : 186412-62-4
  • Molecular Weight : 240.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11018
Availability details for
Quantity
  • Availability
  • Add to Cart