ENA048303861 Enamine

2-(chloromethyl)-5-(4-chlorophenyl)-1,3-oxazole

  • MDL number MFCD04115416
  • Cas Number : 64640-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46-47
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11536
Availability details for
Quantity
  • Availability
  • Add to Cart