ENA048303869 Enamine

5-oxo-1-(1-phenylethyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD02625781
  • Cas Number : 96449-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-202
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11544

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-11544
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart