ENA048303871 Enamine

2-bromo-5-(piperidine-1-sulfonyl)benzoic acid

  • MDL number MFCD00760905
  • Cas Number : 313685-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-143
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11022
Availability details for
Quantity
  • Availability
  • Add to Cart