ENA048303872 Enamine

2-{[(5-acetyl-2-methoxyphenyl)methyl]sulfanyl}acetic acid

  • MDL number MFCD00108110
  • Cas Number : 795290-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11547
Availability details for
Quantity
  • Availability
  • Add to Cart