ENA048303873 Enamine

1-(4-methoxynaphthalen-1-yl)ethan-1-one

  • MDL number MFCD00156696
  • Cas Number : 24764-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-53
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11548
Availability details for
Quantity
  • Availability
  • Add to Cart