ENA048303879 Enamine

2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)-3-methylpentanoic acid

  • MDL number MFCD00430549
  • Cas Number : 19506-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11213

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11213
Availability details for
Quantity
  • Availability
  • Add to Cart