ENA048303892 Enamine

2-bromo-5-[(2-methylpropyl)sulfamoyl]benzoic acid

  • MDL number MFCD06357871
  • Cas Number : 790271-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11028
Availability details for
Quantity
  • Availability
  • Add to Cart