ENA048303893 Enamine

4-{[(oxolan-2-yl)methyl]sulfamoyl}benzoic acid

  • MDL number MFCD01056494
  • Cas Number : 325851-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-170
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11029
Availability details for
Quantity
  • Availability
  • Add to Cart