ENA048303900 Enamine

4-methoxy-N-[(thiophen-2-yl)methyl]aniline

  • MDL number MFCD03043375
  • Cas Number : 3139-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-73
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11575
Availability details for
Quantity
  • Availability
  • Add to Cart