ENA048303901 Enamine

2-{[4-amino-5-(ethoxycarbonyl)pyrimidin-2-yl]sulfanyl}acetic acid

  • MDL number MFCD00435081
  • Cas Number : 115102-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11576
Availability details for
Quantity
  • Availability
  • Add to Cart