ENA048303907 Enamine

2-(5-acetyl-2-methoxyphenyl)acetonitrile

  • MDL number MFCD00119070
  • Cas Number : 403499-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11582
Availability details for
Quantity
  • Availability
  • Add to Cart