ENA048303916 Enamine

2-cyclohexyl-1,3-dioxo-2,3-dihydro-1H-isoindole-5-carboxylic acid

  • MDL number MFCD01046637
  • Cas Number : 67822-74-6
  • Molecular Weight : 273.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-189
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10933
Availability details for
Quantity
  • Availability
  • Add to Cart