ENA048303924 Enamine

2-bromo-5-{[(oxolan-2-yl)methyl]sulfamoyl}benzoic acid

  • MDL number MFCD06655220
  • Cas Number : 857041-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11038
Availability details for
Quantity
  • Availability
  • Add to Cart