ENA048303925 Enamine

2-(2-bromoethyl)-1,2-benzisothiazol-3(2H)-one 1,1-dioxide

  • MDL number MFCD03789713
  • Cas Number : 7248-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11241

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11241
Availability details for
Quantity
  • Availability
  • Add to Cart