ENA048303938 Enamine

4-(phenylamino)-1,2-dihydroquinazoline-2-thione

  • MDL number MFCD02651193
  • Cas Number : 35696-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11251
Availability details for
Quantity
  • Availability
  • Add to Cart