ENA048303944 Enamine

N-(3-chloro-1-phenylpropyl)-4-methylbenzene-1-sulfonamide

  • MDL number MFCD02575343
  • Cas Number : 333429-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11619
Availability details for
Quantity
  • Availability
  • Add to Cart