ENA048303948 Enamine

2-(4-cyclohexylbenzenesulfonamido)acetic acid

  • MDL number MFCD03237555
  • Cas Number : 857041-77-1
  • Molecular Weight : 297.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11046
Availability details for
Quantity
  • Availability
  • Add to Cart