ENA048303951 Enamine

2-{[3-(dimethylsulfamoyl)phenyl]formamido}acetic acid

  • MDL number MFCD03838933
  • Cas Number : 667901-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11626
Availability details for
Quantity
  • Availability
  • Add to Cart