ENA048303969 Enamine

4-[4-(methylsulfanyl)phenyl]-4-oxobutanoic acid

  • MDL number MFCD00082636
  • Cas Number : 7028-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-138
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11051
Availability details for
Quantity
  • Availability
  • Add to Cart