ENA048303973 Enamine

4-methyl-1,3-thiazol-2(3H)-one

  • MDL number MFCD00970121
  • Cas Number : 32497-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11648

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-11648
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart