ENA048303978 Enamine

2-[2-(naphthalen-1-yl)acetamido]propanoic acid

  • MDL number MFCD06347045
  • Cas Number : 150671-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07682
Availability details for
Quantity
  • Availability
  • Add to Cart