ENA048303980 Enamine

3-[(4-chlorobenzyl)thio]-4H-1,2,4-triazol-4-amine

  • MDL number MFCD03484422
  • Cas Number : 522626-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11655

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11655
Availability details for
Quantity
  • Availability
  • Add to Cart