ENA048303984 Enamine

2-cyano-2-cyclopentylideneethanethioamide

  • MDL number MFCD00457737
  • Cas Number : 129224-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11659

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11659
Availability details for
Quantity
  • Availability
  • Add to Cart