ENA048303990 Enamine

1-propyl-2-sulfanyl-1H-1,3-benzodiazole-5-sulfonamide

  • MDL number MFCD04629632
  • Cas Number : 731776-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-193
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11665
Availability details for
Quantity
  • Availability
  • Add to Cart