ENA048303994 Enamine

4-chloro-2-[(4-methylphenyl)amino]-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD03478311
  • Cas Number : 438029-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11286

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11286
Availability details for
Quantity
  • Availability
  • Add to Cart