ENA048303995 Enamine

5-amino-N,2-dimethylbenzene-1-sulfonamide

  • MDL number MFCD06373465
  • Cas Number : 6274-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-135
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11670
Availability details for
Quantity
  • Availability
  • Add to Cart