ENA048303996 Enamine

4-hydrazinothieno[2,3-d]pyrimidine

  • MDL number MFCD00728978
  • Cas Number : 14080-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211...213
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11065

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11065
Availability details for
Quantity
  • Availability
  • Add to Cart