ENA048304003 Enamine

1-benzyl-1H-pyrazol-5-amine

  • MDL number MFCD03596769
  • Cas Number : 3528-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11678

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11678
Availability details for
Quantity
  • Availability
  • Add to Cart