ENA048304005 Enamine

3-(1-Butyl-5-diethylsulfamoyl-1H-benzoimidazol-2-yl)-propionic acid

  • MDL number MFCD03970831
  • Cas Number : 729582-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11680

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11680
Availability details for
Quantity
  • Availability
  • Add to Cart