ENA048304006 Enamine

[2-(3,4-dimethoxyphenyl)ethyl]thiourea

  • MDL number MFCD00041371
  • Cas Number : 21714-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11681
Availability details for
Quantity
  • Availability
  • Add to Cart