ENA048304013 Enamine

2-chloro-N-(2,3-dihydro-1,4-benzodioxin-2-ylmethyl)acetamide

  • MDL number MFCD01250388
  • Cas Number : 119750-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212...214
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11688

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11688
Availability details for
Quantity
  • Availability
  • Add to Cart