ENA048304017 Enamine

3-[(2-methylpiperidin-1-yl)sulfonyl]benzoic acid

  • MDL number MFCD04201336
  • Cas Number : 603118-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11071

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11071
Availability details for
Quantity
  • Availability
  • Add to Cart