ENA048304021 Enamine

2H,3H-imidazo[1,5-a]pyridine-3-thione

  • MDL number MFCD00052489
  • Cas Number : 76259-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11297
Availability details for
Quantity
  • Availability
  • Add to Cart