ENA048304025 Enamine

3-({[(4-methylphenyl)carbamoyl]methyl}sulfanyl)propanoic acid

  • MDL number MFCD06358666
  • Cas Number : 397290-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11076
Availability details for
Quantity
  • Availability
  • Add to Cart