ENA048304036 Enamine

2-(naphthalene-2-sulfonamido)-3-phenylpropanoic acid

  • MDL number MFCD01873085
  • Cas Number : 90427-55-7
  • Molecular Weight : 355.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11308
Availability details for
Quantity
  • Availability
  • Add to Cart