ENA048304037 Enamine

3-(2-oxo-2H-1,4-benzoxazin-3-yl)propanoic acid

  • MDL number MFCD01820402
  • Cas Number : 247571-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11309
Availability details for
Quantity
  • Availability
  • Add to Cart