ENA048304047 Enamine

1-cyclohexyl-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD01128207
  • Cas Number : 313067-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10957
Availability details for
Quantity
  • Availability
  • Add to Cart