ENA048304053 Enamine

(2-phenylethyl)[(thiophen-2-yl)methyl]amine

  • MDL number MFCD04587151
  • Cas Number : 774552-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11728
Availability details for
Quantity
  • Availability
  • Add to Cart