ENA048304195 Enamine

2-(2,4-dimethylphenyl)-5-methyl-2H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD06373471
  • Cas Number : 851175-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11734
Availability details for
Quantity
  • Availability
  • Add to Cart